Merck
  • Protection efficacy of gloves against components of the solvent in a sprayed isocyanate coating utilizing a reciprocating permeation panel.
Protection efficacy of gloves against components of the solvent in a sprayed isocyanate coating utilizing a reciprocating permeation panel.
The Annals of occupational hygiene (2014-12-03)
Diana M Ceballos, Carolyn Reeb-Whitaker, Miyoko Sasakura, Russell Dills, Michael G Yost
ABSTRACT
Determine protection effectiveness of 5-mil natural rubber latex (0.13-mm), 5-mil nitrile rubber (0.13-mm), and 13-mil butyl rubber (0.33-mm) glove materials against solvents present in a commonly used automotive clear coat formulation using a novel permeation panel. The latex and nitrile gloves were the type commonly used by local autobody spray painters. Glove materials were tested by spraying an automotive clear coat onto an automated reciprocating permeation panel (permeation panel II). Temperature, relative humidity, and spray conditions were controlled to optimize clear coat loading homogeneity as evaluated by gravimetric analysis. Solvent permeation was measured using charcoal cloth analyzed by the National Institute for Occupational Safety and Health 1501 method. Natural rubber latex allowed 3-5 times the permeation of solvents relative to nitrile rubber for all 10 solvents evaluated: ethyl benzene, 2-heptanone, 1-methoxy-2-propyl acetate, o-xylene, m-xylene, p-xylene, n-butyl acetate, methyl isobutyl ketone, petroleum distillates, and toluene. There is a distinct behavior in solvent permeation before and after the coating dry time. Solvent permeation increased steadily before coating dry time and remained fairly constant after coating dry time. Butyl was not permeated by any of the solvents under the conditions tested. Commonly used 5-mil thick (0.13-mm) latex and nitrile gloves were ineffective barriers to solvents found in a commonly used clear coat formulation. Conversely, 13-mil (0.33-mm) butyl gloves were found to be protective against all solvents in the clear coat formulation.
Sigma-Aldrich
2-Heptanone, ≥98%, FCC, FG