Merck
Thiazolothiazole-linked porous organic polymers.
Chemical communications (Cambridge, England) (2014-10-21)
Xiang Zhu, Chengcheng Tian, Tian Jin, Jitong Wang, Shannon M Mahurin, Wenwen Mei, Yan Xiong, Jun Hu, Xinliang Feng, Honglai Liu, Sheng Dai
ABSTRACT
Thiazolothiazole-linked porous organic polymers have been synthesized from a facile catalyst-free condensation reaction between aldehydes and dithiooxamide under solvothermal conditions. The resultant porous frameworks exhibit a highly selective uptake of CO2 over N2 under ambient conditions.
Sigma-Aldrich
Dithiooxamide, 97%