Merck
  • Degradation of 4-chloro-3-nitrophenol via a novel intermediate, 4-chlororesorcinol by Pseudomonas sp. JHN.
Degradation of 4-chloro-3-nitrophenol via a novel intermediate, 4-chlororesorcinol by Pseudomonas sp. JHN.
Scientific reports (2014-03-29)
Pankaj Kumar Arora, Alok Srivastava, Vijay Pal Singh
ABSTRACT
A 4-chloro-3-nitrophenol (4C3NP)-mineralizing bacterium, Pseudomonas sp. JHN was isolated from a waste water sample collected from a chemically-contaminated area, India by an enrichment method. Pseudomonas sp. JHN utilized 4C3NP as a sole carbon and energy source and degraded it with the release of stoichiometric amounts of chloride and nitrite ions. Gas chromatography and gas chromatography-mass spectrometry detected 4-chlororesorcinol as a major metabolite of the 4C3NP degradation pathway. Inhibition studies using 2,2'-dipyridyl showed that 4-chlororesorcinol is a terminal aromatic compound in the degradation pathway of 4C3NP. The activity for 4C3NP-monooxygenase was detected in the crude extracts of the 4C3NP-induced JHN cells that confirmed the formation of 4-chlororesorcinol from 4C3NP. The capillary assay showed that Pseudomonas sp. JHN exhibited chemotaxis toward 4C3NP. The bioremediation capability of Pseudomonas sp. JHN was monitored to carry out the microcosm experiments using sterile and non-sterile soils spiked with 4C3NP. Strain JHN degraded 4C3NP in sterile and non-sterile soil with same degradation rates. This is the first report of (i) bacterial degradation and bioremediation of 4C3NP, (ii) formation of 4-chlororesorcinol in the degradation pathway of 4C3NP, (iii) bacterial chemotaxis toward 4C3NP.
Sigma-Aldrich
4-Chloro-3-nitrophenol, 98%