Merck
Ligand effects on Negishi couplings of alkenyl halides.
Organic letters (2011-07-12)
Arkady Krasovskiy, Bruce H Lipshutz
ABSTRACT
Negishi couplings at olefinic centers do not always occur with the anticipated maintenance of stereochemistry. The source of erosion has been traced to the ligand, and a modified method has been developed that solves the stereochemical issue and significantly improves yields of Negishi couplings in general.
Sigma-Aldrich
(2-Bromovinyl)trimethylsilane, approx. 90% trans, 98%