Merck
A high-throughput screening assay for hydroxynitrile lyase activity.
Chemical communications (Cambridge, England) (2006-10-13)
Jennifer Andexer, Jan-Karl Guterl, Martina Pohl, Thorsten Eggert
ABSTRACT
A high-throughput screening assay for hydroxynitrile lyase activity accepting a wide range of HNL-substrates is presented, which is useful either for enzyme fingerprinting or screening of huge variant libraries generated in metagenome or directed evolution approaches.