Merck
  • Solid-phase synthesis of complex oligosaccharides using a novel capping reagent.
Solid-phase synthesis of complex oligosaccharides using a novel capping reagent.
The Journal of organic chemistry (2004-04-03)
Xiangyang Wu, Richard R Schmidt
ABSTRACT
Solid-phase-supported oligosaccharide synthesis of a core N-glycan tetrasaccharide and of a trisaccharide containing the Galili antigen is reported. The synthesis is based on a hydroxymethylbenzyl benzoate spacer-linker system attached to the Merrifield resin, O-Fmoc-protected O-glycosyl trichloroacetimidates as glycosyl donors, and benzoyl isocyanate as a capping reagent for low-reactivity hydroxy groups. In this way, the target molecules could be efficiently obtained with little byproduct formation, and hence final purification was convenient.
Sigma-Aldrich
Benzoyl isocyanate, technical grade, 90%