- Catalytic oxidation of alkanes by iron bispidine complexes and dioxygen: oxygen activation versus autoxidation.
Catalytic oxidation of alkanes by iron bispidine complexes and dioxygen: oxygen activation versus autoxidation.
Chemical communications (Cambridge, England) (2013-11-20)
Peter Comba, Yong-Min Lee, Wonwoo Nam, Arkadius Waleska
PMID24247200
ABSTRACT
Organic substrates (specifically cis-1,2-dimethylcyclohexane, DMCH) are oxidized by O2 in the presence of iron(II)-bispidine complexes. It is shown that this oxidation reaction is not based on O2 activation by the nonheme iron catalysts as in Nature but due to a radical-based initiation, followed by a radical- and ferryl-based catalytic reaction.