Merck
  • Liquid crystals carrying stereodefined vicinal difluoro- and trifluoro- alkyl motifs.
Liquid crystals carrying stereodefined vicinal difluoro- and trifluoro- alkyl motifs.
Chemical communications (Cambridge, England) (2007-12-01)
Marcello Nicoletti, Matthias Bremer, Peer Kirsch, David O'Hagan
ABSTRACT
The synthesis of the stereospecifically fluorinated difluoro- and trifluoro- rac-3 and rac-4 is described where the fluorine atoms are positioned adjacent/vicinal to each other and the physical characteristics of these candidate liquid crystals including negative dielectric anisotropy are measured and rationalised.