Merck
Get up to 22% off for Pi Day until 3/26.Save Now
1-(4-Chloro-benzo-yl)-2,7-dimethoxy-naphthalene.
Acta crystallographica. Section E, Structure reports online (2008-01-01)
Ryosuke Mitsui, Kosuke Nakaema, Keiichi Noguchi, Akiko Okamoto, Noriyuki Yonezawa
ABSTRACT
In the title compound, C(19)H(15)ClO(3), the dihedral angle between the naphthalene ring system and the benzene ring is 72.06 (7)°. The 4-chloro-phenyl group and the carbonyl group are almost coplanar. An inter-molecular C-H⋯O hydrogen bond is formed between an H atom of the 4-chloro-phenyl group and the O atom of one meth-oxy group, forming a zigzag chain along the a axis.