Merck
  • The Drosophila Extradenticle and Homothorax selector proteins control branchless/FGF expression in mesodermal bridge-cells.
The Drosophila Extradenticle and Homothorax selector proteins control branchless/FGF expression in mesodermal bridge-cells.
EMBO reports (2005-07-12)
Samir Merabet, Andreas Ebner, Markus Affolter
ABSTRACT
The stereotyped outgrowth of tubular branches of the Drosophila tracheal system is orchestrated by the local and highly dynamic expression profile of branchless (bnl), which encodes a secreted fibroblast growth factor (FGF)-like molecule. Despite the importance of the spatial and temporal bnl regulation, little is known about the upstream mechanisms that establish its complex expression pattern. Here, we show that the Extradenticle and Homothorax selector proteins control bnl transcription in a single cell per segment, the mesodermal bridge-cell. In addition, we observed that a key determinant of bridge-cell specification, the transcription factor Hunchback, is also required for bnl expression. Therefore, we propose that one of the functions of the bridge-cell is to synthesize and secrete the chemoattractant Bnl. These findings provide a hitherto unknown and interesting link between combinatorial inputs of transcription factors, cell-specific ligand expression and organ morphogenesis.