Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Inhibitors of Abeta production: solid-phase synthesis and SAR of alpha-hydroxycarbonyl derivatives.
Inhibitors of Abeta production: solid-phase synthesis and SAR of alpha-hydroxycarbonyl derivatives.
Bioorganic & medicinal chemistry letters (2003-03-20)
Owen B Wallace, David W Smith, Milind S Deshpande, Craig Polson, Kevin M Felsenstein
ABSTRACT
Inhibitors of amyloid-beta (Abeta) protein production have been widely pursued as a potential treatment for Alzheimer's disease. Following the identification of a 5 microM screening hit, SAR was initiated using solid-phase synthetic techniques. Two series of alpha-hydroxy esters and ketones which are sub-micromolar inhibitors of Abeta production were identified. The most potent alpha-hydroxyketone identified is approximately 30-fold more potent than the initial lead.
Sigma-Aldrich
2,2-Dichloro-1-methyl-cyclopropanecarboxylic acid, 98%