Merck
  • Evaluation of intramitochondrial ATP levels identifies G0/G1 switch gene 2 as a positive regulator of oxidative phosphorylation.
Evaluation of intramitochondrial ATP levels identifies G0/G1 switch gene 2 as a positive regulator of oxidative phosphorylation.
Proceedings of the National Academy of Sciences of the United States of America (2013-12-18)
Hidetaka Kioka, Hisakazu Kato, Makoto Fujikawa, Osamu Tsukamoto, Toshiharu Suzuki, Hiromi Imamura, Atsushi Nakano, Shuichiro Higo, Satoru Yamazaki, Takashi Matsuzaki, Kazuaki Takafuji, Hiroshi Asanuma, Masanori Asakura, Tetsuo Minamino, Yasunori Shintani, Masasuke Yoshida, Hiroyuki Noji, Masafumi Kitakaze, Issei Komuro, Yoshihiro Asano, Seiji Takashima
ABSTRACT
The oxidative phosphorylation (OXPHOS) system generates most of the ATP in respiring cells. ATP-depleting conditions, such as hypoxia, trigger responses that promote ATP production. However, how OXPHOS is regulated during hypoxia has yet to be elucidated. In this study, selective measurement of intramitochondrial ATP levels identified the hypoxia-inducible protein G0/G1 switch gene 2 (G0s2) as a positive regulator of OXPHOS. A mitochondria-targeted, FRET-based ATP biosensor enabled us to assess OXPHOS activity in living cells. Mitochondria-targeted, FRET-based ATP biosensor and ATP production assay in a semiintact cell system revealed that G0s2 increases mitochondrial ATP production. The expression of G0s2 was rapidly and transiently induced by hypoxic stimuli, and G0s2 interacts with OXPHOS complex V (FoF1-ATP synthase). Furthermore, physiological enhancement of G0s2 expression prevented cells from ATP depletion and induced a cellular tolerance for hypoxic stress. These results show that G0s2 positively regulates OXPHOS activity by interacting with FoF1-ATP synthase, which causes an increase in ATP production in response to hypoxic stress and protects cells from a critical energy crisis. These findings contribute to the understanding of a unique stress response to energy depletion. Additionally, this study shows the importance of assessing intramitochondrial ATP levels to evaluate OXPHOS activity in living cells.
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)