Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Glucocorticoid receptor confers resistance to antiandrogens by bypassing androgen receptor blockade.
Glucocorticoid receptor confers resistance to antiandrogens by bypassing androgen receptor blockade.
Cell (2013-12-10)
Vivek K Arora, Emily Schenkein, Rajmohan Murali, Sumit K Subudhi, John Wongvipat, Minna D Balbas, Neel Shah, Ling Cai, Eleni Efstathiou, Chris Logothetis, Deyou Zheng, Charles L Sawyers
ABSTRACT
The treatment of advanced prostate cancer has been transformed by novel antiandrogen therapies such as enzalutamide. Here, we identify induction of glucocorticoid receptor (GR) expression as a common feature of drug-resistant tumors in a credentialed preclinical model, a finding also confirmed in patient samples. GR substituted for the androgen receptor (AR) to activate a similar but distinguishable set of target genes and was necessary for maintenance of the resistant phenotype. The GR agonist dexamethasone was sufficient to confer enzalutamide resistance, whereas a GR antagonist restored sensitivity. Acute AR inhibition resulted in GR upregulation in a subset of prostate cancer cells due to relief of AR-mediated feedback repression of GR expression. These findings establish a mechanism of escape from AR blockade through expansion of cells primed to drive AR target genes via an alternative nuclear receptor upon drug exposure.