Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • The intramolecular rearrangement of phosphinohydrazides [R'2P-NR-NR-M] → [RN═PR'2-NR-M]: general rules and exceptions. transformations of bulky phosphinohydrazines (R-NH-N(PPh2)2, R = tBu, Ph2P).
The intramolecular rearrangement of phosphinohydrazides [R'2P-NR-NR-M] → [RN═PR'2-NR-M]: general rules and exceptions. transformations of bulky phosphinohydrazines (R-NH-N(PPh2)2, R = tBu, Ph2P).
Inorganic chemistry (2012-01-03)
Alexander N Kornev, Vyacheslav V Sushev, Yulia S Panova, Natalia V Belina, Olga V Lukoyanova, Georgy K Fukin, Sergey Y Ketkov, Gleb A Abakumov, Peter Lönnecke, Evamarie Hey-Hawkins
ABSTRACT
Reactions of diphosphinohydrazines R-NH-N(PPh(2))(2) (R = tBu (1), Ph(2)P (3)) with some metalation reagents (Co[N(SiMe(3))(2)](2), LiN(SiMe(3))(2), La[N(SiMe(3))(2)](3), nBuLi, MeLi) were performed. Compound 1 was synthesized by the reaction of Ph(2)PCl with tert-butylhydrazine hydrochloride in 83% yield. This compound reveals temperature-dependent (31)P NMR spectra due to hindered rotation about the P-N bonds. Complicated redox reaction of 1 with Co[N(SiMe(3))(2)](2) proceeds with cleavage of the P-N and N-N bonds to form a binuclear cobalt complex [Co{HN(PPh(2))(2)-κ(2)P,P'}(2)(μ-PPh(2))](2) (2) demonstrating a short Co···Co distance of 2.3857(5) Å, which implies a formal double bond between the Co atoms. Strong nucleophiles (nBuLi, MeLi) cause fragmentation of the molecules 1 and 3, while reactions of 3 with lithium and lanthanum silylamides give products of the NNP → NPN rearrangement [Li{Ph(2)P(NPPh(2))(2)-κ(2)N,N'}(THF)(2)] (4) and [La{Ph(2)P(NPPh(2))(2)-κ(2)N,N'}{N(SiMe(3))(2)}(2)] (5), respectively. These complexes represent the first examples of a κ(2)N,N' bonding mode for the triphosphazenide ligand [(Ph(2)PN)(2)PPh(2)](-). DFT calculations showed large energy gain (52.1 kcal/mol) of the [NNP](-) to [NPN](-) anion rearrangement.
Sigma-Aldrich
tert-Butylhydrazine hydrochloride, 98%