Merck
  • Characterization of polyesters prepared from three different phthalic acid isomers by CID-ESI-FT-ICR and PSD-MALDI-TOF mass spectrometry.
Characterization of polyesters prepared from three different phthalic acid isomers by CID-ESI-FT-ICR and PSD-MALDI-TOF mass spectrometry.
Analytical chemistry (2002-08-30)
Olli Laine, Tuomo Laitinen, Pirjo Vainiotalo
ABSTRACT
Polyesters prepared from the same diol, 2-butyl-2-ethyl-1,3-propanediol, but different phthalic acid isomers, phthalic, isophthalic, and terephthalic acid, were characterized by collision-induced dissociation electrospray ionization Fourier transform ion cyclotron resonance (CID-ESI-FT-ICR) and postsource-decay matrix-assisted laser desorption/ionization time-of-flight (PSD-MALDI-TOF) mass spectrometry. Sodiated dihydroxyl-terminated polyester oligomers containing five repeating units at m/z 1634 were selected as precursor ions for dissociation studies. Two main mechanisms occurred in the fragmentation of all of the polyesters, since dissociation of the oligomers was initiated by hydrogen rearrangement or transesterification reactions. Polyesters prepared from different phthalic acid isomers could be distinguished by their fragmentation behavior. Polyester prepared from phthalic acid was easily identified by using both CID-ESI-FT-ICR and PSD-MALDI-TOF mass spectrometry. However, distinguishing between the polyesters prepared from isophthalic and terephthalic acid succeeded marginally only with CID-ESI-FT-ICR mass spectrometry. Molecular dynamics calculations were used to obtain an idea of the fragmentation behavior of the polyesters. The low-energy structures of the precursor ions were determined, and the coordination of the oxygen atoms of the polyester oligomers to the sodium cation was examined more closely. Both the experimental and the theoretical studies showed that the sodium ion affinity of polyester changed with the phthalic acid isomer.
Sigma-Aldrich
2-Butyl-2-ethyl-1,3-propanediol, 99%