Merck
  • Structure and compatibility of a magnesium electrolyte with a sulphur cathode.
Structure and compatibility of a magnesium electrolyte with a sulphur cathode.
Nature communications (2011-08-11)
Hee Soo Kim, Timothy S Arthur, Gary D Allred, Jaroslav Zajicek, John G Newman, Alexander E Rodnyansky, Allen G Oliver, William C Boggess, John Muldoon
ABSTRACT
Magnesium metal is an ideal rechargeable battery anode material because of its high volumetric energy density, high negative reduction potential and natural abundance. Coupling Mg with high capacity, low-cost cathode materials such as electrophilic sulphur is only possible with a non-nucleophilic electrolyte. Here we show how the crystallization of the electrochemically active species formed from the reaction between hexamethyldisilazide magnesium chloride and aluminum trichloride enables the synthesis of a non-nucleophilic electrolyte. Furthermore, crystallization was essential in the identification of the electroactive species, [Mg(2)(μ-Cl)(3)·6THF](+), and vital to improvements in the voltage stability and coulombic efficiency of the electrolyte. X-ray photoelectron spectroscopy analysis of the sulphur electrode confirmed that the electrochemical conversion between sulphur and magnesium sulfide can be successfully performed using this electrolyte.
Sigma-Aldrich
Hexane, anhydrous, 95%