Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Different G protein-coupled signaling pathways are involved in alpha granule release from human platelets.
Different G protein-coupled signaling pathways are involved in alpha granule release from human platelets.
Journal of thrombosis and haemostasis : JTH (2004-05-14)
T M Quinton, S Murugappan, S Kim, J Jin, S P Kunapuli
ABSTRACT
Alpha granule release plays an important role in propagating a hemostatic response upon platelet activation. We evaluated the ability of various agonists to cause alpha granule release in platelets. Alpha granule release was measured by determining P-selectin surface expression in aspirin-treated washed platelets. ADP-induced P-selectin expression was inhibited both by MRS 2179 (a P2Y1 selective antagonist) and AR-C69931MX (a P2Y12 selective antagonist), suggesting a role for both Galpha(q) and Galpha(i) pathways in ADP-mediated alpha granule release. Consistent with these observations, the combination of serotonin (a Galpha(q) pathway stimulator) and epinephrine (a Galpha(z) pathway stimulator) also caused alpha granule release. Furthermore, U46619-induced P-selectin expression was unaffected by MRS 2179 but was dramatically inhibited by AR-C69931, indicating a dominant role for P2Y12 in U46619-mediated alpha granule release. Additionally, the Galpha(12/13)-stimulating peptide YFLLRNP potentiated alpha granule secretion in combination with either ADP or serotonin/epinephrine costimulation but was unable to induce secretion by itself. Finally, costimulation of the Galpha(i) and Galpha(12/13) pathways resulted in a significant dose-dependent increase in alpha granule release. We conclude that ADP-induced alpha granule release in aspirin-treated platelets occurs through costimulation of Galpha(q) and Galpha(i) signaling pathways. The P2Y12 receptor plays an important role in thromboxane A(2)-mediated alpha granule release, and furthermore activation of Galpha(12/13) and Galpha(q) signaling pathway can cause alpha granule release.
Sigma-Aldrich
YFLLRNP trifluoroacetate, ≥98%