Merck
Agarose gel electrophoresis of denatured RNA with silver staining.
Analytical biochemistry (1988-02-15)
R N Skopp, L C Lane
ABSTRACT
This paper describes agarose gel electrophoresis and silver staining of denatured RNAs. Glyoxal- or formaldehyde-denatured RNAs are electrophoresed in an agarose gel cast on a plastic support using an inert, low conductivity buffer. Following electrophoresis, the gel is stained with a sensitive silver stain. The method produces sharp, well-resolved bands and yields accurate RNA size estimates. Because of its sensitivity and simplicity, it is suitable for routine laboratory use.
Sigma-Aldrich
Glyoxal solution, BioReagent, ~40% in H2O (~8.8 M)