- All conjugated copolymer excitonic multiferroics.
All conjugated copolymer excitonic multiferroics.
Advanced materials (Deerfield Beach, Fla.) (2012-11-23)
Jessica Lohrman, Yueying Liu, Shaofeng Duan, Xiaoyong Zhao, Manfred Wuttig, Shenqiang Ren
PMID23172730
ABSTRACT
A substantial magnetoelectric coupling effect of an excitonic all-conjugated block copolymer multiferroics consisting of electronically distinct polythiophene derivatives is reported. The observations open new avenues for the multifunctional all-conjugated block copolymer synthesis and electric field tunable multiferroic devices.