Merck
  • Cytogenetic effect of the thiocarbamate herbicides butylate, molinate and vernolate in the mouse bone marrow micronucleus test.
Cytogenetic effect of the thiocarbamate herbicides butylate, molinate and vernolate in the mouse bone marrow micronucleus test.
Mutation research (1990-12-01)
A Pintér, M Csík, G Török, A Surján, Z Kelecsényi, Z Kocsis
ABSTRACT
Three thiocarbamate herbicides, butylate (S-ethyl-diisobutylthiocarbamate), vernolate (S-propyl dipropylthiocarbamate) and molinate (S-ethyl-N,N-hexamethylenethiocarbamate) were assayed for cytogenetic effect in the mouse bone marrow micronucleus test. Butylate was inactive in bone marrow, vernolate caused a marginal increase in the incidence of micronucleated polychromatic erythrocytes only at a high toxic dose level. Molinate, the N,N-hexamethylene derivative was, however, strongly active in the bone marrow, causing a high frequency of micronucleated erythrocytes, even at subtoxic concentrations.
Product Description
Supelco
Butylat, PESTANAL®, analytical standard