Merck
  • Antagonism of lateral amygdala alpha1-adrenergic receptors facilitates fear conditioning and long-term potentiation.
Antagonism of lateral amygdala alpha1-adrenergic receptors facilitates fear conditioning and long-term potentiation.
Learning & memory (Cold Spring Harbor, N.Y.) (2010-09-28)
Stephanie C Lazzaro, Mian Hou, Catarina Cunha, Joseph E LeDoux, Christopher K Cain
ABSTRACT
Norepinephrine receptors have been studied in emotion, memory, and attention. However, the role of alpha1-adrenergic receptors in fear conditioning, a major model of emotional learning, is poorly understood. We examined the effect of terazosin, an alpha1-adrenergic receptor antagonist, on cued fear conditioning. Systemic or intra-lateral amygdala terazosin delivered before conditioning enhanced short- and long-term memory. Terazosin delivered after conditioning did not affect consolidation. In vitro, terazosin impaired lateral amygdala inhibitory postsynaptic currents leading to facilitation of excitatory postsynaptic currents and long-term potentiation. Since alpha1 blockers are prescribed for hypertension and post-traumatic stress disorder, these results may have important clinical implications.
Sigma-Aldrich
Terazosin hydrochloride, ≥98% (TLC), powder
Pricing and availability is not currently available.
Terazosin hydrochloride dihydrate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.