Merck
  • Role of prostaglandin D2 receptor DP as a suppressor of tumor hyperpermeability and angiogenesis in vivo.
Role of prostaglandin D2 receptor DP as a suppressor of tumor hyperpermeability and angiogenesis in vivo.
Proceedings of the National Academy of Sciences of the United States of America (2008-12-09)
Takahisa Murata, Michelle I Lin, Kosuke Aritake, Shigeko Matsumoto, Shu Narumiya, Hiroshi Ozaki, Yoshihiro Urade, Masatoshi Hori, William C Sessa
ABSTRACT
Although COX-dependent production of prostaglandins (PGs) is known to be crucial for tumor angiogenesis and growth, the role of PGD(2) remains virtually unknown. Here we show that PGD(2) receptor (DP) deficiency enhances tumor progression accompanied by abnormal vascular expansion. In tumors, angiogenic endothelial cells highly express DP receptor, and its deficiency accelerates vascular leakage and angiogenesis. Administration of a synthetic DP agonist, BW245C, markedly suppresses tumor growth as well as tumor hyperpermeability in WT mice, but not in DP-deficient mice. In a corneal angiogenesis assay and a modified Miles assay, host DP deficiency potentiates angiogenesis and vascular hyperpermeability under COX-2-active situation, whereas exogenous administration of BW245C strongly inhibits both angiogenic properties in WT mice. In an in vitro assay, BW245C does not affect endothelial migration and tube formation, processes that are necessary for angiogenesis; however, it strongly improves endothelial barrier function via an increase in intracellular cAMP production. Our results identify PGD(2)/DP receptor as a new regulator of tumor vascular permeability, indicating DP agonism may be exploited as a potential therapy for the treatment of cancer.