Merck
  • Synthesis of 4-aminotropones from [(sulfinyl or sulfonyl)methyl]-substituted p-quinamines.
Synthesis of 4-aminotropones from [(sulfinyl or sulfonyl)methyl]-substituted p-quinamines.
Chemistry (Weinheim an der Bergstrasse, Germany) (2007-10-09)
M Carmen Carreño, Montserrat Ortega-Guerra, María Ribagorda, M Jesús Sanz-Cuesta
ABSTRACT
An efficient synthesis of 4-aminotropones has been achieved in excellent yields by simple treatment of 4-amino-4-[(p-tolylsulfinyl)methyl]-2,5-cyclohexadienones (p-quinamines) with NaH. The method allowed regiocontrolled access to 3-methyl, 5-methyl- and 3,5-dimethyl-substituted derivatives starting from p-quinamines with adequate substituents at the cyclohexadienone moiety and/or at the carbon linked to the sulfur function. The p-quinamines in turn were easily accessible from N-Boc p-anisidines (Boc=tert-butoxycarbonyl) by electrochemical oxidation in MeOH to quinone imine monoketals, followed by addition of a alpha-lithium sulfinyl carbanion to the imino group, and ketal hydrolysis. Oxidation of the sulfoxide gave the sulfonyl-substituted p-quinamines that, upon basic treatment, behave similarly. The p-quinamine 55 and bis-p-quinamine 56, resulting in the addition of the anion derived from dimethyl sulfone to the p-quinonimine ketal 14, also gave the 4-aminotropone. The mechanism involves the initial formation of a alpha-sulfonyl carbanion, which intramolecularly attacks the cyclohexadienone giving a norcaradiene-like enolate intermediate, the evolution of which through a ring-expansion process, pushes off a methyl sulfinate anion or SO2. This efficient process fulfils the criteria of atom economy. The introduction of a proline substituent in the nitrogen of the starting p-quinamine allowed the synthesis of an enantiopure 4-aminotropone, the asymmetric Diels-Alder reactions of which with maleimide occurred in a highly endo and pi-facial diastereoselective manner.
Sigma-Aldrich
Tropone, 97%