Merck
  • Drug transport mechanisms in helminth parasites: passive diffusion of benzimidazole anthelmintics.
Drug transport mechanisms in helminth parasites: passive diffusion of benzimidazole anthelmintics.
Experimental parasitology (2006-01-25)
L Mottier, L Alvarez, L Ceballos, C Lanusse
ABSTRACT
Anthelmintic molecules must reach their receptors inside target parasites to exert the pharmacological effect. Available data suggest that the main route of entry of antiparasitic drugs into helminth parasites would be through their external surface. However, it is unclear if trans-tegumental/cuticular penetration is the most important way of entry of benzimidazole (BZD) anthelmintics into their target parasites compared to oral ingestion. The relative involvement of active and passive transport mechanisms has not been defined. The goal of the work reported here was to determine the main processes involved in the entry of BZD anthelmintic molecules into the three main classes of helminth parasites. Adult specimens of Moniezia benedeni (cestode), Fasciola hepatica (trematode) and Ascaris suum (nematode) were incubated in Kreb's Ringer Tris buffer (pH 7.4, 37 degrees C) (1g parasite/10 ml incubation medium) for 15, 45, and 90 min, respectively, in the presence of a concentration gradient of either fenbendazole (FBZ), oxfendazole or triclabendazole sulphoxide (TCBZSO) (1-30 mol/ml, n=4). Dead helminth specimens were also incubated with the same drug concentration gradient. Specimens of F. hepatica with the oral route closed off by ligation were incubated with TCBZSO in the presence or absence of bovine serum albumin. After the incubation time elapsed, samples of parasite material were chemically extracted and prepared for high performance liquid chromatography analysis to measure drug/metabolite concentrations. Equivalent drug concentrations were measured within ligated and non-ligated liver flukes, demonstrating that BZD do mainly penetrate by trans-tegumental diffusion. The higher the concentration of BZD molecules in the incubation medium, the greater their concentration recovered within the helminth parasites. High correlation coefficients (>0.98) were obtained between initial drug concentration in the incubation medium and those measured inside the nematode, cestode, and trematode parasites. FBZ concentrations recovered from tissues of dead cestodes/nematodes over time were significantly greater compared to those measured in living parasites. These differences in drug diffusion may be related to the morphological/functional properties of the parasite's external surfaces. The outcome of the work reported here indicates that passive drug transfer through the external helminth surface is the main transport mechanism accounting for BZD accumulation into target parasites.
Product Description
Supelco
Triclabendazole sulfoxide, VETRANAL®, analytical standard