Merck
  • Formulation development of inhalation powders for FK888 with carrier lactose using Spinhaler and its absorption in healthy volunteers.
Formulation development of inhalation powders for FK888 with carrier lactose using Spinhaler and its absorption in healthy volunteers.
Journal of controlled release : official journal of the Controlled Release Society (2004-05-19)
Toshiomi Nakate, Hiromitsu Yoshida, Atsuo Ohike, Yuji Tokunaga, Rinta Ibuki, Yoshiaki Kawashima
ABSTRACT
(4R)-4-Hydroxy-l-[(l-methyl-lH-indol-3-yl)carbonyl]-L-prolyl-N-benzyl-N-methyl-3-(2-naphthyl)-L-alaninamide (FK888) is a candidate selective NK1 receptor antagonist, and it exhibits poor absorption from the gastrointestinal (GI) tract in healthy volunteers. The objective of this study was to develop an optimized DPI formulation with carrier lactose using a Spinhaler, and thereby improve the systemic absorption of FK888. The fine particles of FK888 were blended with various carrier lactoses, and in vitro deposition properties were investigated using a twin impinger. The mixture using 100 M and 325 M lactoses [Sieved lactoses (SLs)] exhibited a higher emitted dose (Em) than 200 M, 450 M and micronized lactoses [Milled lactoses (MLs)]. The flowability of carrier lactose had an influence on the Em. On the other hand, the respirable particle (RP) fraction in the formulations with MLs was much higher than that of SLs, in spite of the blended ratios of lactose. It was also observed that the mixture of 325 M with the micronized lactose particles had the same RP as 200 M, although the 325 M alone had a low RP. Considering the Em and RP obtained, we chose 200 M for FK888 dry powder inhaler (DPI). The proportional absorption was found up to the 12.5% of the FK888 ratio (5 mg as unit dose) for the Cmax and AUC in healthy volunteers. In conclusion, 200 M, which has fine lactose particles and a better flowability than other MLs, is an extremely suitable carrier for maximizing the fine particle dose as far as FK888 is concerned. Furthermore, an improvement in the systemic absorption of FK888 was achieved using the dry powder formulations.