Merck
  • Spectrophotometric estimation of D-penicillamine in bulk and dosage forms using 2,6-dichloroquinone-4-chlorimide (DCQ).
Spectrophotometric estimation of D-penicillamine in bulk and dosage forms using 2,6-dichloroquinone-4-chlorimide (DCQ).
Journal of pharmaceutical and biomedical analysis (2000-03-04)
A A Al-Majed
ABSTRACT
A simple colorimetric method for the determination of D-penicillamine in pure form and pharmaceutical formulations is described. The method is based on coupling between D-penicillamine and 2,6-dichloroquinone-4-chlorimide (DCQ) in dimethylsulphoxide. The optimum conditions for the reaction were investigated and incorporated into the procedure. The reaction forms a yellow 1:1 complex with maximum absorbance at 431 nm (epsilon = 3,700). Regression analysis of Beer's plot showed good correlation (r = 0.9998) and the calibration graph was rectilinear over the range 4-20 microg/ml(-1) with a detection limit of 0.15 microg/ml(-1) . The average recovery for the commercial capsules was 101.66% with an RSD of 1.57%. The results obtained were sufficiently accurate, reproducible and in accordance with those given by the official method. The method is specific for the intact drug, and can be adopted in the presence of some interfering substances.
Sigma-Aldrich
6-(2,2-Dicyanovinyl)-N-(2-hydroxyethyl)-1,2,3,4-tetrahydroquinoline