Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Formation of omeprazole sulphone but not 5-hydroxyomeprazole is inhibited by grapefruit juice.
Formation of omeprazole sulphone but not 5-hydroxyomeprazole is inhibited by grapefruit juice.
British journal of clinical pharmacology (2000-02-15)
W Tassaneeyakul, S Vannaprasaht, Y Yamazoe
ABSTRACT
To determine the effect of grapefruit juice on omeprazole metabolism in vivo. This was a randomized crossover study with a 2 week washout period. Omeprazole (20 mg) was taken orally by 13 healthy volunteers after an overnight fast with either grapefruit juice or water. Serial blood samples were obtained over 12 h and standardized meals were served 3 and 10 h after the administration of omeprazole. Plasma concentrations of omeprazole and its major metabolites, 5-hydroxyomeprazole and omeprazole sulphone, were determined by high performance liquid chromatography (h.p.l.c.). Mean area under the plasma concentration vs time curve (AUC) between 0 and 12 h for omeprazole sulphone was approximately 20% lower (P<0.01) in the group receiving grapefruit juice. There was no significant difference in the mean AUC of 5-hydroxyomeprazole or omeprazole. The AUC ratio of omeprazole sulphone to omeprazole, an index of CYP3A4 activity, was decreased by 33% (P<0.001) after administration of grapefruit juice whereas the AUC ratio of 5-hydroxyomeprazole to omeprazole, an index of CYP2C19 activity, did not differ between the two phases of the study. Although the time to peak concentration (tmax ) and terminal half-life (t1/2,z) of omeprazole and its two main metabolites were not altered, the peak concentration (Cmax ) of omeprazole sulphone was significantly reduced after administration of grapefruit juice. Administration of grapefruit juice decreased the formation of omeprazole sulphone but not 5-hydroxyomeprazole. These results indicate that activities of CYP3A4, but not of CYP2C19, are inhibited by the simultaneous administration of grapefruit juice.
Product Description
Omeprazole impurity D, European Pharmacopoeia (EP) Reference Standard