- Catalytic asymmetric sulfenylation of unprotected 3-substituted oxindoles.
Catalytic asymmetric sulfenylation of unprotected 3-substituted oxindoles.
Organic letters (2012-05-17)
Yunfei Cai, Jun Li, Weiliang Chen, Mingsheng Xie, Xiaohua Liu, Lili Lin, Xiaoming Feng
PMID22587589
ABSTRACT
Catalytic asymmetric sulfenylation of unprotected 3-substituted oxindoles has been developed via cooperative catalysis of a chiral N,N'-dioxide-Sc(OTf)(3) complex and a Brønsted base. Utilizing readily available N-(phenylthio)phthalimide as the sulfur source, a wide range of optically active 3-phenylthiooxindoles were obtained in excellent yields with excellent enantioselectivities under mild reaction conditions.