Merck
  • [The reaction of 3-(R-phenyl)-6-hydrazine pyridazine with 5-methylisatin and 1-morpholinomethyl-5-methylisatin].
[The reaction of 3-(R-phenyl)-6-hydrazine pyridazine with 5-methylisatin and 1-morpholinomethyl-5-methylisatin].
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi (1996-01-01)
M Dorneanu, E Stefănescu, O Păduraru, M Pavelescu, A Hriscu
ABSTRACT
This paper presents the synthesis of six hydrazones obtained by treating 5-methyl-isatin or 1-morpholino methyl-5-methyl-isatin with 3-(R-phenyl)-6-hydrazino-pyridazine (R = OCH3, Cl, Br) and two complex combination with copper, derived from 3-(p-anisyl-pyridazinyl)-hydrazone-5-methyl-indoline-2- one. The structure of the new compounds was confirmed by the results of the quantitative elementary and IR, UV-VIS spectral analysis. The biological tests point out that product VI, in which a copper atom binds two molecules of 3-(p-anisyl-pyridazinyl)-hydrazono-5-methyl-indoline-2-one, has a considerable antiinflammatory activity, giving a inflammation inhibition of 39, 6%. All the synthetized compounds have a moderate antimicrobial activity against Candida albicans.
Sigma-Aldrich
5-Methylisatin, 95%