Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • The multiple-sugar metabolism (msm) gene cluster of Streptococcus mutans is transcribed as a single operon.
The multiple-sugar metabolism (msm) gene cluster of Streptococcus mutans is transcribed as a single operon.
FEMS microbiology letters (1996-07-01)
R E McLaughlin, J J Ferretti
ABSTRACT
The multiple-sugar metabolism (msm) locus of Streptococcus mutans constitutes a non-PTS sugar uptake system responsible for the transport and utilization of raffinose, melibiose and isomaltotrioses. While previous studies have used polar mutations to suggest that these genes are co-transcribed, there has not been evidence to support this. In this report we present direct evidence that the msm genes can be transcribed as a single operon.