Merck
  • [Combination of antimycotic preparations and natural proteinase inhibitors in otomycosis].
[Combination of antimycotic preparations and natural proteinase inhibitors in otomycosis].
Revue de laryngologie - otologie - rhinologie (1993-01-01)
Y A Soushko, O N Borissenko, L I Volosevitch
ABSTRACT
In vitro experiments, Terrilytinum* increases the antimycotic activity of Clotrimazolum* and Nitrofungin*, and cancels out the inhibitory effect of Contrycal*. The combined application of Clotrimazolum and Nitrofungin with Terrilytinum and Contrycal as treatment of otomycosis of the middle ear has an outstanding clinical and anti-mycotic efficacy. The results of the investigation show that the proposed method of treatment is marked by an excellent therapeutic efficacy in the case of otomycosis.
Sigma-Aldrich
2-Chloro-4-nitrophenol, 97%