Merck
  • Metabolic detoxication of bis-(2-hydroxy-3, 5-dichlorophenyl)-sulfoxide in man.
Metabolic detoxication of bis-(2-hydroxy-3, 5-dichlorophenyl)-sulfoxide in man.
The Journal of toxicological sciences (1981-11-01)
M Sakamoto, K Chiba, I Agata, T Higa, M Kiyota, N Hayashi, N Yagi, M Itoh, H Sekikawa, M Takada
ABSTRACT
The metabolic detoxication of bis(2-hydroxy-3, 5-dichlorophenyl)sulfoxide (BTS) in man was investigated. Bis(2-hydroxy-3, 5-dichlorophenyl)sulfide (BT) was identified in beta-glucuronidase treated urine following the administration of BTS by thin-layer chromatography, gas chromatography, ultraviolet spectrum and quantitative analysis. No other metabolites were detectable. BT-glucuronide was also identified in urine. It was assumed that BTS was reduced to BT and successively conjugated with glucuronide in man, and excreted as BT-glucuronide in the urine.