Merck
The sensitizing potential of 2-ethylhexyl acrylate in the guinea pig.
Contact dermatitis (1983-09-01)
T H Waegemaekers, H B van der Walle
ABSTRACT
The sensitizing potential of 2-ethylhexyl acrylate in the guinea pig could be demonstrated by Freund's Complete Adjuvant Test. This acrylate ester is a common constituent of adhesive tape. Allergic reactions to several brands of adhesive tape were not observed in 2-ethylhexyl acrylate sensitized animals. Cross reactions with other acrylic monomers were observed.
Product Description
Supelco
2-Ethylhexyl acrylate, analytical standard
Sigma-Aldrich
2-Ethylhexyl acrylate, 98%, contains ≥0.001-≤0.11% monomethyl ether hydroquinone as stabilizer