Merck
[Hypolactatemic effect of sodium difluoroacetate].
Comptes rendus des seances de la Societe de biologie et de ses filiales (1980-01-01)
J M Dalstein, G Ribes, P Campo, M M Loubatières-Mariani
ABSTRACT
In the anesthetized rat, the intraperitoneal injection of 40 mg/kg sodium difluoroacetate (DFA), an activator of the pyruvate dehydrogenase, counteracted the hyperlactatemia induced by a high dose of phenformin (40 mg/kg) injected concomitantly. In the normal conscious dog, the administration of 150 mg/kg by gastric intubation decreased the blood lactate and pyruvate levels; however, this effect was less marked than that produced by the same dose of sodium dichloroacetate (DCA).
Sigma-Aldrich
Difluoroacetic acid, 98%