Merck
  • Reaction of nucleic acids bases and their derivatives with peroxodisulfate ion.
Reaction of nucleic acids bases and their derivatives with peroxodisulfate ion.
Nucleic acids symposium series (1985-01-01)
T Itahara, R Ebihara, Y Seto, Y Fujii, M Tada
ABSTRACT
Reaction with peroxodisulfate ion was investigated, that is, reaction of 1,3-dimethyluracil, 1,3-dimethylthymine, and caffeine with carbon radicals formed from decarboxylation of carboxylic acids, oxidation of the methyl group at 5-position of thymines, and halogenation of nucleic acids bases and their derivatives with alkali halides.
Sigma-Aldrich
1,3-Dimethyluracil, 99%