Merck
  • Pantothenases from pseudomonads produce either pantoyl lactone or pantoic acid.
Pantothenases from pseudomonads produce either pantoyl lactone or pantoic acid.
The Biochemical journal (1988-03-01)
R K Airas
ABSTRACT
A pseudomonad was separated having a pantothenase that produced beta-alanine and pantoic acid. The pantothenase from an old strain, Pseudomonas fluorescens UK-1, was shown to produce beta-alanine and pantoyl lactone. The pantoic acid-producing pantothenase was characterized and compared with the pantothenase from the strain UK-1. The Mr was 240,000; it apparently consists of four subunits. The Km value for pantothenate is 3 mM. The enzymic activity is affected by an ionizable group of pK 8.4, the enzyme is active at higher pH, and V but not Km is affected by pH. This pantothenase is not inhibited by di-isopropyl phosphorofluoridate or phenylmethanesulphonyl fluoride, unlike the enzyme from the strain UK-1. Both pantothenases are inhibited by m-aminophenylboronic acid, oxalate, oxaloacetate and Cl- ions. The pantoic acid-producing pantothenase is inhibited also by SO4(2-) ions. The strong inhibitions by many compounds make this pantothenase unsuitable for the assay of pantothenic acid.
Sigma-Aldrich
(S)-(+)-Pantolactone, 97%
Sigma-Aldrich
DL-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, purum, ≥97.0% (T)
Sigma-Aldrich
D-(−)-Pantolactone, 99%