Merck
  • Low-dose propranolol and exercise capacity in postural tachycardia syndrome: a randomized study.
Low-dose propranolol and exercise capacity in postural tachycardia syndrome: a randomized study.
Neurology (2013-04-26)
Amy C Arnold, Luis E Okamoto, André Diedrich, Sachin Y Paranjape, Satish R Raj, Italo Biaggioni, Alfredo Gamboa
ABSTRACT
To determine the effect of low-dose propranolol on maximal exercise capacity in patients with postural tachycardia syndrome (POTS). We compared the effect of placebo vs a single low dose of propranolol (20 mg) on peak oxygen consumption (VO2max), an established measure of exercise capacity, in 11 patients with POTS and 7 healthy subjects in a randomized, double-blind study. Subjects exercised on a semirecumbent bicycle, with increasing intervals of resistance to maximal effort. Maximal exercise capacity was similar between groups following placebo. Low-dose propranolol improved VO2max in patients with POTS (24.5 ± 0.7 placebo vs 27.6 ± 1.0 mL/min/kg propranolol; p = 0.024), but not healthy subjects. The increase in VO2max in POTS was associated with attenuated peak heart rate responses (142 ± 8 propranolol vs 165 ± 4 bpm placebo; p = 0.005) and improved stroke volume (81 ± 4 propranolol vs 67 ± 3 mL placebo; p = 0.013). In a separate cohort of POTS patients, neither high-dose propranolol (80 mg) nor metoprolol (100 mg) improved VO2max, despite similar lowering of heart rate. These findings suggest that nonselective β-blockade with propranolol, when used at the low doses frequently used for treatment of POTS, may provide a modest beneficial effect to improve heart rate control and exercise capacity. This study provides Class II evidence that a single low dose of propranolol (20 mg) as compared with placebo is useful in increasing maximum exercise capacity measured 1 hour after medication.
Product Description
Propranolol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Propranolol hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder