Merck
  • Open-tubular electrochromatographic chiral separation of amino acids using an organic nanocrystals immobilized capillary.
Open-tubular electrochromatographic chiral separation of amino acids using an organic nanocrystals immobilized capillary.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2013-01-11)
Fumihiko Kitagawa, Hiroshi Sudaki, Kenji Sueyoshi, Koji Otsuka
ABSTRACT
The preparation of cinchonidine (CCND) nanocrystals and their immobilization onto the inner surface of a fused silica capillary was investigated for use in the enantioseparation by capillary electrochromatography. The CCND nanocrystals were prepared by an emulsion method that utilizes crystal growth in an oil-in-water emulsion without a stabilizer. As a result, white-colored aqueous dispersions of organic nanoparticles were obtained. SEM images showed that the prepared CCND nanocrystals were from 300 to 700 nm in size. When the obtained dispersion was introduced into a poly(diallyldimethylammonium chloride) (PDDAC) coated capillary, the nanocrystals were strongly adsorbed onto the inner surface due to an electrostatic interaction between cationic PDDAC and the negatively charged organic nanocrystals. In CCND nanocrystals coated capillary, CEC enantioseparations of racemic amino acids were successfully achieved. The reproducibilities of the detection times were acceptable with a relative standard deviation of less than 7%, indicating that stable nanocrystal coating could be obtained by our proposed method.
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw <100,000 (very low molecular weight), 35 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 200,000-350,000 (medium molecular weight), 20 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, 20 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 400,000-500,000 (high molecular weight), 20 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cinchonidine, 96%