Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Treatment of Aroclor 1016 contaminated soil by hydrogen peroxide: laboratory column study.
Treatment of Aroclor 1016 contaminated soil by hydrogen peroxide: laboratory column study.
Environmental technology (2012-12-18)
Marika Viisimaa, Jelena Veressinina, Anna Goi
ABSTRACT
The potential and feasibility of treating soil contaminated with electrical insulating oil, Aroclor 1016, containing polychlorinated biphenyls (PCBs) with stabilized hydrogen peroxide were evaluated using columns packed with soils of two different matrixes. The column experiments showed that PCBs degraded by the stabilized hydrogen peroxide treatment in both soil matrixes, although the efficacy of the treatment depended strongly on the soil characteristics. The removal of PCB-containing oil was higher in sandy silt soil than in sandy soil. While a higher iron content promoted hydrogen peroxide oxidation of the contaminant in sandy silt soil, lower permeability and higher organic matter content contributed to an oxidation decrease as a function of depth. Dehydrogenase activity measurements indicated no substantial changes in microbial activity during the treatment of both sandy and sandy silt soils, thus offering opportunities to apply the hydrogen peroxide treatment to the remediation of PCB-contaminated soil.
Product Description
Supelco
Aroclor 1016 solution, certified reference material, 1000 μg/mL in isooctane