Merck
  • Safety and efficacy of the cobalt chromium PRO-Kinetik coronary stent system: results of the MULTIBENE study.
Safety and efficacy of the cobalt chromium PRO-Kinetik coronary stent system: results of the MULTIBENE study.
Cardiovascular revascularization medicine : including molecular interventions (2012-11-21)
Paul Vermeersch, Yolande Appelman, Dieter Horstkotte, Gert Richardt, Jean Boland, Jacques Lalmand, Patrick Coussement, Marc Castadot, Luc Janssens, Pierfrancesco Agostoni, Ian Buysschaert, Maarten J Suttorp
ABSTRACT
The aim of this MULTIBENE study was to evaluate the safety and efficacy of the silicon carbide coated cobalt chromium PRO-Kinetik coronary stent system in patients with single de novo coronary lesions. This prospective international multicenter study included 202 patients at 10 European sites. Analysis was performed on the per protocol population of 197 patients. Patients were followed until 12 months, a subset of patients (n=72) underwent additional coronary angiography at 6 months. Primary endpoint was 6-months rate of target vessel failure (TVF), defined as a composite of cardiac death, myocardial infarction and target vessel revascularization (TVR). At 6 months, rate of TVF was 10.9% and rate of major adverse cardiac events, a composite of cardiac death, MI, target lesion revascularization (TLR) and coronary artery bypass graft, was 11.4%, both being mainly attributed to TVR respective TLR. No cardiac death or stent thrombosis occurred. In-segment late lumen loss was 0.66±0.61mm and binary restenosis was 20.8%, as determined by core laboratory in the angiographic subgroup. Based on these data, the PRO-Kinetik coronary stent system was found to be safe and effective.
Sigma-Aldrich
Silicon carbide, nanopowder, <100 nm particle size
Sigma-Aldrich
Silicon carbide, −400 mesh particle size, ≥97.5%
Sigma-Aldrich
Silicon carbide, -200 mesh particle size