Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • A terbium(III)-organic framework for highly selective sensing of cytidine triphosphate.
A terbium(III)-organic framework for highly selective sensing of cytidine triphosphate.
The Analyst (2012-10-04)
Xi Juan Zhao, Rong Xing He, Yuan Fang Li
ABSTRACT
Highly selective sensing of cytidine triphosphate (CTP) against other triphosphate nucleosides including ATP, GTP and UTP is successfully achieved with a luminescent terbium(III)-organic framework (TbOF) of [Tb(2)(2,3-pzdc)(2)(ox)(H(2)O)(2)](n) (2,3-pzdc(2-) = 2,3-pyrazinedicarboxylate, ox(2-) = oxalate).