Merck
  • Traceless directing group for stereospecific nickel-catalyzed alkyl-alkyl cross-coupling reactions.
Traceless directing group for stereospecific nickel-catalyzed alkyl-alkyl cross-coupling reactions.
Organic letters (2012-05-10)
Margaret A Greene, Ivelina M Yonova, Florence J Williams, Elizabeth R Jarvo
ABSTRACT
Stereospecific nickel-catalyzed cross-coupling reactions of benzylic 2-methoxyethyl ethers are reported for the preparation of enantioenriched 1,1-diarylethanes. The 2-methoxyethyl ether serves as a traceless directing group that accelerates cross-coupling. Chelation of magnesium ions is proposed to activate the benzylic C-O bond for oxidative addition.
Sigma-Aldrich
Ethane, 99.99%
Sigma-Aldrich
Ethane, ≥99%