Merck
  • Phosphine-catalyzed domino reaction for the synthesis of conjugated 2,3-dihydrofurans from allenoates and Nazarov reagents.
Phosphine-catalyzed domino reaction for the synthesis of conjugated 2,3-dihydrofurans from allenoates and Nazarov reagents.
Chemistry, an Asian journal (2012-04-19)
Peizhong Xie, Wenqing Lai, Zhishuai Geng, You Huang, Ruyu Chen
ABSTRACT
A new domino reaction for Nazarov reagents: An efficient approach was developed for the construction of highly functionalized conjugated 2,3-dihydrofuran skeletons. Nazarov reagents were used for the first time in a phosphine-catalyzed domino reaction and successfully used to construct five-membered ring compounds using alcohol as the solvent. DFT calculations indicate that alcohol is essential for the catalysis of the [1,2]-proton transfer.
Sigma-Aldrich
2,3-Dihydrofuran, 99%