Merck
  • Detection of melamine and cyanuric acid in vegetable protein products used in food production.
Detection of melamine and cyanuric acid in vegetable protein products used in food production.
Journal of food science (2012-03-16)
Lawrence R Levinson, Kimberley A Gilbride
ABSTRACT
The multitude of food recalls in 2007 clearly demonstrated that total nitrogen-content (ΣN) determination by means of near-infrared spectroscopy (NIRS) and Kjeldahl-based measurements can be deceived, and should no longer be regarded as a complete quality assurance program for nutritive-protein evaluations. Furthermore, contemporary Canadian-employed analytical tools are precariously limited in their ability to effectively assure a product where there is no a priori knowledge of the environmental toxin(s) involved. In light of these challenges, this study explored a number of analytical techniques used to assess and furthermore assure the quality of vegetable protein products (VPPs). Using liquid chromatography with tandem mass spectrometry (LC/MS/MS) technologies, a combination of VPP-based samples was analyzed for the presence of nitrogen-bearing environmental toxicants. Of the 52 samples tested, involving an assortment of matrices, melamine and cyanuric acid were positively identified (>1 ng/mL) in 22 and 17 samples, respectively. Subsequent high pressure liquid chromatography with ultraviolet/visible (HPLC-UV) amino acid profiling further confirmed the adulteration of those materials contaminated with melamine and melamine-related compounds. Based on the evidence presented herein, LC/MS/MS in combination with HPLC-UV provides for a reliable food safety detection system as applied to VPPs. Moreover, HPLC-UV is indispensable as a stand-alone 1st level of screening to assess the integrity of a VPP or any nutritive protein-based sample.
Product Description
Supelco
Cyanuric acid, analytical standard
Sigma-Aldrich
Cyanuric acid, 98%