Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Efficacy of plant-derived and synthetic compounds on clothing as repellents against Ixodes scapularis and Amblyomma americanum (Acari: Ixodidae).
Efficacy of plant-derived and synthetic compounds on clothing as repellents against Ixodes scapularis and Amblyomma americanum (Acari: Ixodidae).
Journal of medical entomology (2012-02-09)
Robert A Jordan, Terry L Schulze, Marc C Dolan
ABSTRACT
We conducted field trials to compare the relative repellent activity of two natural product compounds (nootkatone and carvacrol) with commercially available plant-derived (EcoSMART organic insect repellent) and permethrin-based (Repel Permanone) repellents against adult Ixodes scapularis Say and Amblyomma americanum (L.) (Acari: Ixodidae) by using treated coveralls. One day after treatment, nootkatone and carvacrol provided 100% repellency of I. scapularis adults, with nootkatone maintaining complete protection through 3 d, whereas carvacrol showed steadily declining repellency against I. scapularis during the 7-d course of the trials. Nootkatone was at least as effective against host-seeking A. americanum as against I. scapularis through 3 d. Carvacrol provided little protection against A. americanum adults. Both natural compounds performed well initially in comparison with the commercial products. After 7 d, nootkatone was the most effective against both species followed in order of activity by Permanone, EcoSMART, and carvacrol. Nootkatone seems to have offer considerable potential as a clothing repellent against both I. scapularis and A. americanum.
Sigma-Aldrich
(+)-Nootkatone, ≥99.0% (GC)