Merck
  • Reaction of acetals with various carbon nucleophiles under non-acidic conditions: C-C bond formation via a pyridinium-type salt.
Reaction of acetals with various carbon nucleophiles under non-acidic conditions: C-C bond formation via a pyridinium-type salt.
Chemistry, an Asian journal (2011-12-14)
Hiromichi Fujioka, Kenzo Yahata, Tomohito Hamada, Ozora Kubo, Takashi Okitsu, Yoshinari Sawama, Takuya Ohnaka, Tomohiro Maegawa, Yasuyuki Kita
ABSTRACT
Mild substitution reactions of acetals with carbon nucleophiles via the pyridinium-type salts generated by the treatment of acetals with TESOTf-2,4,6-collidine or 2,2'-bipyridyl have been developed. Various carbon nucleophiles, such as organocuprates, silyl enol ethers, enamines, etc., reacted with the pyridinium-type salts to give the corresponding substituted products in good yields. The reactions proceeded under very mild conditions (non-acidic conditions) and thus acid-sensitive functional groups can be tolerated during the reaction. In addition, only an acetal can form the pyridinium-type salt and react with nucleophiles in the presence of a ketal. This unusual selectivity is in contrast to general methods conducted under acidic conditions.
Sigma-Aldrich
2,4,6-Trimethylpyridine, puriss. p.a., 99% (GC)
Sigma-Aldrich
2,4,6-Trimethylpyridine, ReagentPlus®, 99%