Merck
  • Probing the origin of degenerate metathesis selectivity via characterization and dynamics of ruthenacyclobutanes containing variable NHCs.
Probing the origin of degenerate metathesis selectivity via characterization and dynamics of ruthenacyclobutanes containing variable NHCs.
Journal of the American Chemical Society (2011-09-17)
Benjamin K Keitz, Robert H Grubbs
ABSTRACT
The preparation of new phosphonium alkylidene ruthenium metathesis catalysts containing N-heterocyclic carbenes (NHCs) that result in a preference for degenerate metathesis is described. The reaction of the catalysts with ethylene or substrates relevant to ring-closing metathesis (RCM) produced ruthenacyclobutanes that could be characterized by cryogenic NMR spectroscopy. The rate of α/β methylene exchange in ethylene-only ruthenacycles was found to vary widely between ruthenacycles, in some cases being as low as 3.97 s(-1) at -30 °C, suggesting that the NHC plays an important role in degenerative metathesis reactions. Attempts to generate RCM-relevant ruthenacycles resulted in the low-yielding formation of a previously unobserved species, which we assign to be a β-alkyl-substituted ruthenacycle. Kinetic investigations of the RCM-relevant ruthenacycles in the presence of excess ethylene revealed a large increase in the kinetic barrier of the rate-limiting dissociation of the cyclopentene RCM product compared with previously investigated catalysts. Taken together, these results shed light on the degenerate/productive selectivity differences observed for different metathesis catalysts.
Sigma-Aldrich
Butane, 99%