Merck
  • Redox-based thrombelastographic method to detect carboxyhemefibrinogen-mediated hypercoagulability.
Redox-based thrombelastographic method to detect carboxyhemefibrinogen-mediated hypercoagulability.
Blood coagulation & fibrinolysis : an international journal in haemostasis and thrombosis (2011-08-09)
Vance G Nielsen, Matthew R Arkebauer, Keith Vosseller
ABSTRACT
Cigarette smoking is associated with plasmatic hypercoagulability, and carbon monoxide has been demonstrated to enhance coagulation by binding to a fibrinogen-bound heme. Our objective was to design and test a redox-based method to detect carboxyhemefibrinogen. Normal, pooled, citrated plasma was exposed to 0-100  μmol/l carbon monoxide releasing molecule-2 (tricarbonyldichlororuthenium (II) dimer; CORM-2) before or after exposure to the organic reductant phenylhydroxylamine (PHA, 0-30  mmol/l), a compound that rapidly converts Fe(+2) to Fe(+3) in heme. Addition of calcium and tissue factor activation in disposable thrombelastographic cups was performed, followed by data collection at 37°C for 15  min. Elastic modulus (G, dynes/cm(2)) was the primary endpoint. CORM-2 significantly increased G values by 67.8% compared to unexposed plasma; pretreatment with 10  mmol/l PHA significantly decreased G values in CORM-2-exposed plasma by 77.1%, whereas 30  mmol/l PHA was required to significantly decrease G values by 64.0% in plasma following CORM-2 pre-exposure. G values were not significantly different between unexposed plasma and plasma exposed to CORM-2 followed by 30  mmol/l PHA addition. Conversion of fibrinogen-bound to the metheme state alone decreased G by 34.3-38.9% following exposure to 10-30  mmol/l PHA. Conversion of fibrinogen-bound heme Fe(+2) to Fe(+3) with PHA abrogated carbon monoxide-mediated increases in clot strength. Clinical trials are planned to investigate smoking individuals to mechanistically link carboxyhemefibrinogen formation with in-vitro hypercoagulability.
Sigma-Aldrich
N-Phenylhydroxylamine, ≥95.0%