Merck
  • An efficient route to 1-aminoisoquinolines via AgOTf-catalyzed reaction of 2-alkynylbenzaldoxime with amine.
An efficient route to 1-aminoisoquinolines via AgOTf-catalyzed reaction of 2-alkynylbenzaldoxime with amine.
Organic & biomolecular chemistry (2011-05-28)
Danqing Zheng, Zhiyuan Chen, Jianping Liu, Jie Wu
ABSTRACT
2-Alkynylbenzaldoxime reacts with amine catalyzed by silver triflate under mild conditions, leading to 1-aminoisoquinolines in good yield. This reaction proceeds efficiently with good functional group tolerance.
Sigma-Aldrich
1-Aminoisoquinoline, 99%
Pricing and availability is not currently available.