Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Bepridil facilitates early termination of spiral-wave reentry in two-dimensional cardiac muscle through an increase of intercellular electrical coupling.
Bepridil facilitates early termination of spiral-wave reentry in two-dimensional cardiac muscle through an increase of intercellular electrical coupling.
Journal of pharmacological sciences (2010-12-16)
Hiroki Takanari, Haruo Honjo, Yoshio Takemoto, Tomoyuki Suzuki, Sara Kato, Masahide Harada, Yusuke Okuno, Takashi Ashihara, Tobias Opthof, Ichiro Sakuma, Kaichiro Kamiya, Itsuo Kodama
ABSTRACT
Bepridil is effective for conversion of atrial fibrillation to sinus rhythm and in the treatment of drug-refractory ventricular tachyarrhythmias. We investigated the effects of bepridil on electrophysiological properties and spiral-wave (SW) reentry in a 2-dimensional ventricular muscle layer of isolated rabbit hearts by optical mapping. Ventricular tachycardia (VT) induced in the presence of bepridil (1 µM) terminated earlier than in the control. Bepridil increased action potential duration (APD) by 5% - 8% under constant pacing and significantly increased the space constant. There was a linear relationship between the wavefront curvature (κ) and local conduction velocity: LCV = LCV₀ - D·κ (D, diffusion coefficient; LCV₀, LCV at κ = 0). Bepridil significantly increased D and LCV₀. The regression lines with and without bepridil crossed at κ = 20 - 40 cm⁻¹, resulting in a paradoxical decrease of LCV at κ > 40 cm⁻¹. Dye transfer assay in cultured rat cardiomyocytes confirmed that bepridil increased intercellular coupling. SW reentry in the presence of bepridil was characterized by decremental conduction near the rotation center, prominent drift, and self-termination by collision with boundaries. These results indicate that bepridil causes an increase of intercellular coupling and a moderate APD prolongation, and this combination compromises wavefront propagation near the rotation center of SW reentry, leading to its drift and early termination.
Sigma-Aldrich
Bepridil hydrochloride, powder